0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Gold
Gold ≥99.95% (metals basis), wire, annealed, Ø 0.025 mm (0.001 in)
Gold ≥99.95% (metals basis), wire, annealed, Ø 0.025 mm (0.001 in)
:
Thermo Fisher Scientific
Gold ≥99.95% (metals basis), wire, annealed, Ø 0.025 mm (0.001 in)
:
Thermo Fisher Scientific
:
7440-57-5
:
:
Au
:
MFCD00003436
:
196.97 g/mol
:
12,04526
:
2800 °C (1013 hPa)
:
1063 °C
:
19.3 g/cm³ (20 °C)
:
Ambient
:
231-165-9
msds_ALFA040585
Thermo Scientific Gold Products
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals