TE buffer, 1X, Molecular Biology Grade, Ultrapure
Use directly to re-suspend and/or dilute purified DNA or RNA.
- 1X TE solution consisting of 10 mM Tris, and 1 mM EDTA
- Solution is 0,2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles
- Environmentally Preferable:
Frequently Bought Together