Order Entry
Czech Republic
ContactUsLinkComponent
Erbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Flammable
Erbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Catalog # 12255.14
CAS Number:  
Flammable
Erbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Catalog # 12255.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :167.26 g/mol
ADR:4.1,II
 :Packaged under argon.
EINECS:231-160-1
UN:3178
 :GHS02
Storage Temperature:Ambient
CAS Number:7440-52-0
Formula:Er
MDL Number:MFCD00010987