

Cerium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-45-1
:
Formula:Ce
MDL Number:MFCD00010924
:140.12 g/mol
Boiling Pt:3468 °C (1013 hPa)
Melting Pt:795 °C
Density:∼6.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-154-9
UN:1333
ADR:4.1,II
:GHS02
:GHS07
Frequently Bought Together