Tin ≥99.98% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-31-5
:
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.310
:118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
Frequently Bought Together