ContactUsLinkComponent
Tin ≥99.995% (metals basis), slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 2 mm (0.1 in)
Tin ≥99.995% (metals basis), slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 2 mm (0.1 in)
  41169.KD
 :  
Tin ≥99.995% (metals basis), slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 2 mm (0.1 in)
  41169.KD
 :  
  • Pk:
    50
  • Seq:
    0050
 :7440-31-5
 :
 :Sn
 :MFCD00133862
 :12,09587
 :2270 °C (1013 hPa)
 :232 °C
 :7.31 g/cm³ (25 °C)
 :118.71 g/mol
 :Ambient
 :231-141-8