Tin ≥99.999% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Catalog # 00344.GE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1727
- Pk Info:75×100 mm
Specifications
About this item
:
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
:118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8
CAS Number:7440-31-5
Formula:Sn
MDL Number:MFCD00133862