Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Charcoal activated
Charcoal activated, slug 6-8 mesh
Charcoal activated, slug 6-8 mesh
:
Thermo Fisher Scientific
Charcoal activated, slug 6-8 mesh
:
Thermo Fisher Scientific
:
7440-44-0
:
64365-11-3
:
C
:
MFCD00133992
:
12.01 g/mol
:
13,01818
:
3550 °C
:
2 g/cm³ (20 °C)
:
Ambient
:
231-153-3
:
1362
:
4.2,III
:
01-2119488894-16
msds_ALFA088764
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
: Merck
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
: VWR Chemicals
2-Propanol ≥99.9% (by GC), LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck