Order Entry
Czech Republic
ContactUsLinkComponent
Indium ≥99.99% (metals basis), ingot/button, Ø ∼31 mm (1.2 in), Thickness: 5.3 mm (0.21 in)
Indium ≥99.99% (metals basis), ingot/button, Ø ∼31 mm (1.2 in), Thickness: 5.3 mm (0.21 in)
Catalog # 45111.KF
CAS Number:  
Indium ≥99.99% (metals basis), ingot/button, Ø ∼31 mm (1.2 in), Thickness: 5.3 mm (0.21 in)
Catalog # 45111.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-74-6
 :
Formula:In
MDL Number:MFCD00134048
 :114.82 g/mol
Merck Index:13,04971
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:156 °C
Density:7.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-180-0