
Gallium ≥99.99999% (metals basis), ingot
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-55-3
:
Formula:Ga
MDL Number:MFCD00134045
:69.72 g/mol
Merck Index:12,04364
Boiling Pt:2403 °C (1013 hPa)
Melting Pt:29 °C
Density:5.904 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:231-163-8
UN:2803
ADR:8,III
:GHS05
Related Information
Frequently Bought Together