Order Entry
Czech Republic
ContactUsLinkComponent
Aluminium ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Aluminium ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40829.KS
CAS Number:  
Aluminium ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40829.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7429-90-5
 :
Formula:Al
MDL Number:MFCD00134029
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3