Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness: 1.0 µm
Catalog # 40684.DJ
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mm
- Seq:1667
Specifications
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-072-3