Magnesium, turnings for grignard-reaction, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7439-95-4
:
Formula:Mg
MDL Number:MFCD00085308
:24.3 g/mol
:M-Clarity™ quality level = MQ200
EINECS:231-104-6
:GHS02
Storage Temperature:Ambient
Frequently Bought Together