Cerium(IV) oxide 20% in water, colloidal dispersion
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1306-38-3
:
Formula:CeO₂
MDL Number:MFCD00010933
:172.11 g/mol
Density:1.22
Storage Temperature:Ambient
EINECS:215-150-4
Related Information
Frequently Bought Together