Sodium nitrite 0.1 M NaNO2 (0.1N) volumetric solution, Fluka™
Catalog # 35273-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:69 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118