Capillary columns, traditional phases, SP-2380, Supelco®
: Merck
These columns are situated between the traditional non bonded cyanosilicone phases SP-2330 and SP-2340 in polarity. The high polarity of this phase allows the separation of geometric (cis/trans) fatty acid methyl ester isomers as a group.
- Temperature limit from subambient to 275 °C
: Meets USP G48 requirements.
Frequently Bought Together