Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Polydimethylsiloxane
Polydimethylsiloxane, 10 cSt, Sigma-Aldrich®
Polydimethylsiloxane, 10 cSt, Sigma-Aldrich®
:
Merck
Polydimethylsiloxane, 10 cSt, Sigma-Aldrich®
:
Merck
:
63148-62-9
:
9016-00-6
:
(CH₃)₃SiO(Si(CH₃)₂O)n
:
MFCD00084411
:
>140 °C (0.002 mmHg)) (lit.)
:
0.93 g/ml (25 °C)
:
M-Clarity™ quality level = MQ100
Frequently Bought Together
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol, LiChrosolv®, gradient grade for liquid chromatography, Supelco®
: Merck
Poly(methyl hydrogen siloxane) for synthesis, Sigma-Aldrich®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
: Merck
Dimethyl sulphoxide, SupraSolv® for headspace gas chromatography, Supelco®
: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals