Order Entry
Czech Republic
ContactUsLinkComponent
Nephrotoxicity library, Screen-Well®
Nephrotoxicity library, Screen-Well®
Catalog # ENZOENZLIB1000100
Supplier:  Enzo Life Sciences
CAS Number:  
Nephrotoxicity library, Screen-Well®
Catalog # ENZOENZLIB1000100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Description:
    Screen-Well® Nephrotoxicity library, 100 µl/well
  • Seq:
    02
  • Concentration:
    Solvent and concentration are specified in the documentation provided with the library

Specifications

About this item

The Screen-Well® nephrotoxicity library is a focused collection of 86 compounds with defined and diverse nephrotoxicity, including analgesic nephropathy, interstitial nephritis, glomerularopathy, proximal and distal tubulopathy, acute renal failure, chronic kidney disease, and more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. Most of the compounds are dissolved in DMSO at 10 mM unless otherwise stated in the documentation provided with the library. Selected compounds are dissolved in DI water. All compounds are provided at 100 μl volumes in a 96-well plate. This library is an essential cost-effective tool for predictive toxicology screening and assay development.

The Screen-Well® Nephrotoxicity Library is a focused collection of 86 compounds with defined and diverse nephrotoxicity, including analgesic nephropathy, interstitial nephritis, glomerularopathy, proximal and distal tubulopathy, acute renal failure, chronic kidney disease, and more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. Most of the compounds are dissolved in DMSO at 10mM unless otherwise stated in the documentation provided with the library. Selected compounds are dissolved in DI water. All compounds are provided at 100µL volumes in a 96-Well plate. This library is an essential cost-effective tool for predictive toxicology screening and assay development.

Ordering information: Contains 86 compounds with defined and diverse nephrotoxicity. A variety of structurally and mechanistically different compound classes, as well as non toxic controls, are included.