N-Acetyl-Leu-Leu-Methional
Supplier: Enzo Life Sciences
Certificates
About this item
Calpain inhibitor
Cell-permeable, peptide aldehyde inhibitor of calpain and other neutral cysteine proteases. Potent inhibitor of cathepsin L (Ki=0,6 nM) and cathepsin B (Ki=100 nM).
Specifications
- Protein/peptide name:N-Acetyl-Leu-Leu-Methional
- Purity:95% (HPLC)