0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteins and Peptides
PKI
PKI
ENZOBMLP2040001
:
ENZO LIFE SCIENCES
:
PKI
ENZOBMLP2040001
:
ENZO LIFE SCIENCES
:
Pk:
1 mg
Seq:
02
Protein/peptide name:
PKI
Purity:
97%
PKA inhibitor
Competitive inhibitor of PKA (Ki =1,7 nM).