Autocamtide-2
Katalogové číslo ENZOBMLP1010001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:02
- Protein/peptide name:Autocamtide-2
- Purity:>95% (HPLC)
- Formulation:Lyophilised
Specifications
About this item
CaM kinase inhibitor
Sequence, KKALRRQETVDAL, derived from Ca2+/calmodulin-dependent protein kinase II autophosphorylation site RQETVD plus basic residues to facilitate phosphocellulose binding. Contains consensus phosphorylation sequence R-X-X-S/T. Highly selective CaMKII substrate (Km=2 µM for CaMKII; 50 µM for PKC; no activity towards PKA).