Order Entry
Czech Republic
ContactUsLinkComponent
Nuclear extract from HeLa cells
Nuclear extract from HeLa cells
Catalog # ENZOBMLKI1400100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Nuclear extract from HeLa cells
Catalog # ENZOBMLKI1400100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µl
  • Description:
    Nuclear extractor
  • Seq:
    02
  • Storage buffer:
    Liquid. In 0,1 M KCl, 20 mM HEPES/NaOH, pH 7,9, 20% (v/v) glycerol, 0,2 mM EDTA, 0,5 mM DTT, 0,5 mM PMSF.

Specifications

About this item

Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0,5 µl or less per well is sufficient for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.