Ac-IETD-CHO
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
Caspase and granzyme inhibitor
Reversible inhibitor of caspase-6, -8 and granzyme B. Also inhibits caspase-10.
Specifications
- Protein/peptide name:Ac-IETD-CHO
- Purity:>97% (HPLC)
- Formulation:Lyophilized
Specifications
Frequently Bought Together