Liver (human) microsome extract
Catalog # ENZOADILYTHM100F
Supplier: Enzo Life Sciences
Specifications
- Description:Human liver microsome extract
- Seq:02
- Storage conditions:−80 °C
- Application:Western blot
- Pk:200 µG
Specifications
About this item
Microsomes are a rich source of heme-containing metabolic enzymes, including heme-oxygenases and cytochrome P450 (CYP) enzymes, and are commonly used in the study of xenobiotic drug metabolism. Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells.
- Preparation pooled from various human donors of mixed genders
Liquid extract in a SDS-PAGE sample buffer (pH: 8,8) containing 40 mM TRIS-hydrogen chloride, 1% SDS, 50 mM DTT, 7,5% glycerol and 0,003% bromophenol blue.