Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant FGF-8
  ENZOENZPRT1740025
 :  
Human recombinant FGF-8
  ENZOENZPRT1740025
 :  

 

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein synonyms:
    Fibroblast growth factor-8|AIGF|FGF-8b|HBGF-8
  • Protein/peptide name:
    FGF-8
  • Purity:
    >95% (SDS and HPLC)
  • Molecular weight:
    ~22,5 kDa
  • Formulation:
    Lyophilized from 10 mM sodium phosphate, pH 8,0, containing 75 mM NaCl.
  • Purification:
    Sterile filtered through a 0,2 micron filter.

 

 

Produced in E. coli.