Human recombinant FGF-4
Catalog # ENZOENZPRT1730025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:Transforming protein KS3|HST-1|HBGF-4|Fibroblast growth factor-4
- Protein/peptide name:FGF-4
- Purity:>95% (SDS and HPLC)
- Molecular weight:~19,7 kDa
- Formulation:Lyophilized from 10 mM sodium phosphate, pH 7,4, containing 100 mM NaCl.
- Purification:Sterile filtered through a 0,2 micron filter.
Specifications
About this item
Produced in E. coli.