Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant Lin28a (full-length)
Human recombinant Lin28a (full-length)
Supplier:  Enzo Life Sciences
Certificates
Human recombinant Lin28a (full-length)
Supplier:  Enzo Life Sciences

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    microRNA|cancer|Protein lin-28 homolog A|stem cells
  • Protein/peptide name:
    Lin28a
  • Purity:
    >95% (SDS-PAGE)
  • Molecular weight:
    ~24 kDa
  • Formulation:
    Liquid. In 20 mM TRIS, pH 7,2, containing 200 mM sodium chloride and 5 mM diothiothreitol (DTT).
  • Purification:
    Purified by multi-step chromatography.

Specifications

Frequently Bought Together