Human recombinant eNOS
Catalog # ENZOALX2018530010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Protein synonyms:Nitric oxide synthase (endothelial)|NOS III|Endothelial NOS
- Protein/peptide name:eNOS
- Purity:>90% (SDS-PAGE, Western Blot)
- Molecular weight:~135kDa/subunit; homodimer
- Formulation:Liquid. In 50 mM TRIS, pH 7,4, containing 0,1 mM EDTA, 10% glycerol, 100 mM NaCl, 0,1 mM DTT, 10 µM tetrahydrobiopterin.
Specifications
About this item
Produced in Sf9 cells. Untagged human eNOS.