Mouse recombinant FGF basic
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. Non-glycosylated protein, containing 145 amino acids.
- Carrier-free
Specifications
- Protein/peptide type:Recombinant
- Species:Mouse
- Biological activity:The activity is determined by the dose-dependent proliferation of BALB/3T3 cells which is typically observed at concentations <1 ng/ml.
- Protein synonyms:Prostatropin|HBGF-2|FGF2
- Protein/peptide name:FGF basic
- Purity:>95% (Reducing and Non-reducing SDS-PAGE)
- Molecular weight:~16,3kDa
- Formulation:Lyophilized from 5 mM Na2PO4, pH 7,5 and 50 mM sodium chloride. Sterile filtered.
Specifications
Frequently Bought Together