Mouse recombinant IL-6
Catalog # ENZOALX2018190010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Mouse
- Seq:02
- Biological activity:The activity is determined by dose-dependent induction of 7TD1 cell proliferation which is typically observed at concentations <0,02 ng/ml.
- Protein synonyms:Interleukin-6|HSF|BSF-2|IFN-beta2|B-cell differentiation factor|MGI-2
- Protein/peptide name:IL-6
- Purity:>95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
- Molecular weight:~21,7kDa
- Formulation:Lyophilized. Sterile filtered.
Specifications
About this item
Produced in E. coli. Non-glycosylated protein, containing 187 amino acids.
- Carrier-free