Human recombinant NF-kappaB p65 subunit (12-317) (GST-tag) (from E. coli), GST tag
Catalog # ENZOBMLUW99900050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:NF-kappaB p65 subunit (12-317) (GST-tag)
Specifications
About this item
Produced in E. coli.