Human recombinant USP15 (from E. coli), HIS tag
Catalog # ENZOBMLUW98450100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:USP15
Specifications
About this item
Produced in E. coli.