Human Recombinant ISG15 (from E. coli)
Catalog # ENZOBMLUW92350500
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:ISG15
Specifications
About this item
Produced in E. coli.
Mature protein for functional studies.