Mouse recombinant [R112A, R113G]brain derived neurotrophic factor precursor (from E. coli)
Catalog # ENZOBMLSE5480005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Seq:03
- Protein/peptide name:[R112A, R113G]Brain derived neurotrophic factor precursor
- Purity:>99%
Specifications
About this item
Trk ligand
Produced in E. coli.