Human Recombinant 14-3-3zeta (from E. coli), HIS Tag
Catalog # ENZOBMLSE4850100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:14-3 -3 zeta
- Purity:>90% (SDS-PAGE)
Specifications
About this item
Produced in E. coli. Human 14-3-3ζ is fused at the C-terminus to a His-tag.