Human Recombinant 14-3-3gamma (from E. coli), HIS Tag
Catalog # ENZOBMLSE4820100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:HIS Tag
- Protein/peptide name:14-3 -3 gamma
- Purity:>85% (SDS-PAGE)
Specifications
About this item
Produced in E. coli. Human 14-3-3gamma is fused at the C-terminus to a His-tag.