Human recombinant PKA C-alpha (GST-tag),GST tag
Catalog # ENZOBMLSE4100005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:PKA C-alpha (GST-tag)
Specifications
About this item
Produced in insect cells. Full length active enzyme fused at the N-terminus to a GST-tag.