Human Recombinant 14-3-3zeta (GST-tag) (from E. coli), GST Tag
Catalog # ENZOBMLSE3180100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Tag sequence:GST Tag
- Protein/peptide name:14-3 -3 zeta (GST-tag)
Specifications
About this item
Produced in E. coli. Human 14-3-3ζ is fused at the N-terminus to a GST-tag.