Human recombinant GCN5 (from E. coli)
Catalog # ENZOBMLSE2720050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Protein/peptide name:GCN5
- Purity:>95% (SDS-PAGE)
- Concentration:0,5 mg/ml
- Formulation:Liquid. In 20 mM sodium citrate, pH 6,0, containing 0,25M NaCl, 1 mg/ml BSA and 10% glycerol.
Specifications
About this item
Produced in E. coli. Catalytic domain (aa 497-663) of human hGCN5L.