Order Entry
Czech Republic
ContactUsLinkComponent
 
Human recombinant proteasome activator 11S gamma subunit
Catalog # ENZOBMLPW98750100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant proteasome activator 11S gamma subunit
Catalog # ENZOBMLPW98750100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Seq:
    02
  • Protein/peptide name:
    Proteasome activator 11S gamma subunit

Specifications

About this item