mLCK Substrate (from Skeletal and smooth muscle)
Catalog # ENZOBMLP1140001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Source:Skeletal and smooth Muscle
- Seq:02
- Protein/peptide name:mLCK Substrate
- Purity:97%
Specifications
About this item
This sequence, KKRAARATSDVFA, is chicken gizzard smooth muscle myosin P-light chain (MLC 11 to 23) with Pro to Ala14 and Gln to Ala15 substitutions.
Contains specificity determinants for MLC phosphorylation (target is Ser19) and is an excellent myosin light chain kinase (MLCK) substrate. Km=8,6 µM for rabbit skeletal muscle MLCK, 7,5 µM for chicken gizzard smooth muscle MLCK.