Tetanus toxoid
Catalog # ENZOALX630108C100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:100 µG
- Seq:02
- Reconstitution instructions:Reconstitute with 200 µl distilled water to get a 0,5 µg/µl solution in 100 mM histidine, 100 mM sodium chloride and 2% trehalose, pH 7,0,
- Protein/peptide name:Tetanus toxoid
- Concentration:0,5 µg/µl after reconstitution
- Formulation:Lyophilised
Specifications
About this item
Inactivation with formaldehyde leads to non-toxic form of tetanus toxin.
Produced from tetanus toxin by inactivation with formaldehyde. The resulting toxoid is essentially non-toxic.