Human recombinant omentin 1
ENZOALX522108C010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 100 µl sterile water. PBS containing at least 0,1% BSA should be used for further dilutions.
- Protein/peptide name:Omentin 1
- Purity:>95% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Produced in HEK 293 cells. Human omentin 1 (intelectin-1) (aa 17-313) is fused at the N-terminus to a FLAG®-tag.
- Increases AKT phosphorylation in the absence and presence of insulin
- May play a role in the defense system against microorganisms
- May be involved in iron metabolism
Has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes.
May specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner