Human recombinant TL1A (soluble) (from E. coli)
Catalog # ENZOALX522084C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Seq:02
- Reconstitution instructions:Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
- Protein/peptide name:TL1A (soluble)
- Purity:>85% (SDS-PAGE)
- Concentration:0,1 mg/ml after reconstitution
- Formulation:Lyophilised. Contains PBS.
Specifications
About this item
Produced in E. coli. The extracellular domain of human TL1A (aa 93-251) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.