Human recombinant LAG-3:Fc (from CHO cells)
ENZOALX522078C050
:
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Seq:02
- Biological activity:Inhibits binding of MAb to LAG-3 (human) to LAG-3. Induces maturation of human dendritic cells.
- Protein/peptide name:LAG-3:Fc
- Purity:>99% (SDS-PAGE)
- Concentration:0,5 mg/ml
- Formulation:Liquid. In PBS.
Produced in CHO cells. The sequence coding for the 4 extracellular Ig-like domains of human LAG-3 (D1-D4) is fused to the Fc portion of human IgG1.