Order Entry
Czech Republic
ContactUsLinkComponent
Human;Mouse recombinant Fc:CD40L (soluble)
Human;Mouse recombinant Fc:CD40L (soluble)
Catalog # ENZOALX522072C010
Supplier:  Enzo Life Sciences
CAS Number:  
Human;Mouse recombinant Fc:CD40L (soluble)
Catalog # ENZOALX522072C010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
    Mouse
  • Seq:
    02
  • Biological activity:
    Stimulates the proliferation of mouse B cells and dendritic cells. Note: Does not require an enhancer.
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Protein/peptide name:
    Fc:CD40L (soluble)
  • Purity:
    >90% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to the Fc portion of human IgG1 and a linker peptide (20 aa).