Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant BAFF (soluble) (from E. coli)
Human recombinant BAFF (soluble) (from E. coli)
Catalog # ENZOALX522025C010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant BAFF (soluble) (from E. coli)
Catalog # ENZOALX522025C010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Seq:
    02
  • Biological activity:
    Mediates splenocyte survival. Induces monocyte survival.
  • Reconstitution instructions:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    BAFF (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in E. coli. The extracellular domain of human BAFF (aa 83-285) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.