Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant SuperFasLigand™ (soluble)
Human recombinant SuperFasLigand™ (soluble)
Supplier:  Enzo Life Sciences
Certificates
undefined
Human recombinant SuperFasLigand™ (soluble)
Supplier:  Enzo Life Sciences

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Biological activity:
    Kills Fas-sensitive cells at concentrations of >1 ng/ml.
    Note: Does not require enhancer.
  • Reconstitution instructions:
    Reconstitute with 50 µl sterile water. Further dilutions should be made with cell culture medium containing 5% fetal calf serum.
  • Protein/peptide name:
    SuperFasLigand™ (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0,1 mg/ml after reconstitution
  • Formulation:
    Lyophilised. Contains PBS.

Specifications

Frequently Bought Together