Mouse nerve growth factor 25S
Catalog # ENZOALX521006C010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 µG
- Species:Mouse
- Seq:02
- Biological activity:mNGF 2.5S was bioassayed for neurotrophic activity using the rat pheochromocytoma cell PC12 cultures over a period of 7 - 14 days. The neurotrophic activity is in the range of 0,5 - 50 ng/ml. The recommended concentration to be used in vitro for maintenance
- Reconstitution instructions:Reconstitute in sterile water or DMEM at a concentration of at least 10 µg/ ml and store at –80 °C before use. Medium with mNGF cannot be reused due to mNGF degradation.
- Protein/peptide name:Nerve Growth Factor 25S
- Purity:>98% (SDS-PAGE)
- Formulation:Lyophilised
Specifications
About this item
Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984).
Ordering information: Quantity: mNGF 2.5S concentration is estimated spectroscopically from its extinction coefficient and amino acid analysis.