Zadání objednávky
Czech Republic
ContactUsLinkComponent
Human recombinant TNF alpha
Human recombinant TNF alpha
  • Typ proteinu/peptidu:
    Recombinant
  • Druhy:
    Human
  • Biologická aktivita:
    Induces cytolysis of mouse L929 cells in the presence of 1µg/ml Actinomycin D (ED₅₀≤0,05 ng/ml).
  • Pokyny pro rekonstituce:
    A quick spin is recommended followed by reconstitution in distilled, sterile water to a concentration of 0,1 - 1 mg/ ml. For further dilution, add a carrier protein to avoid loss of biological activity.
  • Název proteinu/peptidu:
    TNF alpha
  • Čistota:
    >97% (SDS-PAGE)
  • Formulace:
    Lyophilised from 0,2 μm-filtered solution containing mannitol.
  • Třída:
    Cell culture Grade
Frequently Bought Together